Selenium AA standards, BAKER INSTRA-ANALYZED®, J.T. Baker®
6465-04EA 19578 INR
BAKR6465-04
Selenium AA standards, BAKER INSTRA-ANALYZED®, J.T. Baker®
The standard is prepared by the careful dilution of the metal or its salt, usually 99,99% pure as verified by arc emission spectrometry
  • Standardised with NIST standard chemicals
  • Maximum shelf life and stability
The final concentration of each lot is determined by titrimetry or gravimetry.