Lead AA standard, BAKER INSTRA-ANALYZED®, J.T. Baker®
6455-04EA 26308 INR
BAKR6455-04
Lead AA standard, BAKER INSTRA-ANALYZED®, J.T. Baker®
lead standard prepared by dissolution of lead metal of 99,999% spectral purity. The metal content of the solution is confirmed by titration with EDTA, traceable to NIST SRM 915.
  • Standardised with NIST standard chemicals