Thymol blue alcoholic solution, BAKER ANALYZED® PharmaTest™ USP Test Solution (TS), J.T. Baker®

Supplier: Avantor
Danger

Synonyms: Thymolsulphonphthalein

5929-01EA 10152.16 INR
5929-01
Thymol blue alcoholic solution, BAKER ANALYZED® PharmaTest™ USP Test Solution (TS), J.T. Baker®
Thymol blue
Formula: C₂₇H₃₀O₅S
MW: 466,6 g/mol
Density: 0,83 g/cm³
Flash Pt: 13,00 °C
MDL Number: MFCD00005869
CAS Number: 76-61-9
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Visual Transition Interval - pHÖ9.6 (Blue) Passes Test
Visual Transition Interval - pHÖ1.2 (Red) Passes Test
Visual Transition Interval - pHÖ2.8 (Yellow) Passes Test
Visual Transition Interval - pHÖ8.0 (Yellow) Passes Test
Meets test solution (T.S.) specifications for testing USP/NF monographs.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR