Aluminum, 1000 µg/ml (0,10% w/v), BAKER INSTRA-ANALYZED® reagent
5701-04EA 0 INR
BAKR5701-04
Aluminum, 1000 µg/ml (0,10% w/v), BAKER INSTRA-ANALYZED® reagent
Plasma standard.
  • Prepared from the highest purity raw material 99,999%
  • Assay value accurate within ±0,3% of the assay value
  • Certificate of analysis lists the trace impurity level in µg/ml for over 65 elements