Poly-L-lysine slides, Chromaview™
Supplier: Thermo Fisher Scientific
Chromaview™
RUNC47100EA 0 INR
RUNC47100
Poly-L-lysine slides, Chromaview™
Slides Adhesive Slides
White glass microscope slides coated in poly-L-lysine to promote adhesion.
Designed for use in immunohistology applications and advanced staining techniques.