Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nicotinic acid

Supplier: U.S. Pharmacopeia
Warning



Formula: C₆H₅NO₂
MW: 123,11 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 142 °C
Density: 1,473 g/cm³ (20 °C)
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 200 mg USPH1461003
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty