Adenine ≥98.0%, high purity

Danger

Synonyms: 7H-Purin-6-amine6-AminopurinePurin-6-amine

0183-50GEA 6394 INR
0183-50G
Adenine ≥98.0%, high purity
Adenine

A purine derivative with essential functions for all known life forms. Required for energy metabolism and as a component of DNA and RNA. Component of many culture media.

Formula: C₅H₅N₅
MW: 135,13 g/mol
Density: 0,99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00152

Order Now

Specification Test Results

Heavy Metals ≤0.001%
Identification (IR) PASS
Loss on Drying 1.00%
Nitrogen Content 50.2 – 53.4%
Purity 98.00%
Residue on Ignition 0.10%
Solubility (0.05%, Water) NONE

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR