Iron(II) sulphate hydrate 86-89% (by anhydrous basis) USP
|
Warning
|
Synonyms:
Ferrous sulphate hydrate
E990-500GEA
4187
INR
E990-500G
Iron(II) sulphate hydrate 86-89% (by anhydrous basis) USP
Iron (II) sulphate hydrate
Formula:
FeSO₄·H₂O MW: 169,93 g/mol Boiling Pt: >300 °C Melting Pt: 57 °C Density: 2,841 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149722 CAS Number: 13463-43-9 EINECS: 231-753-5 Merck Index: 12,04105 |
Specification Test Results
Arsenic |
|
Insolubles | ≤0.05% |
Lead | ≤0.001% |
Mercury | ≤3 ug/g |
Purity (Anhydrous) | 86 – 89% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...