Zinc chloride ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
|
Danger
|
Management of Change (MOC) category = N
4322-01EA
21927.08
INR
4322-01
Zinc chloride ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Zinc chloride
Formula:
ZnCl₂ MW: 136,3 g/mol Boiling Pt: 732 °C (1013 hPa) Melting Pt: 283 °C Density: 2,91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 12,10261 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Store at 2 to 25 °C | |
ACS - Assay (ZnCl₂) | ≥ 97.0 % |
ACS - Oxychloride | Passes Test |
ACS - Insoluble Matter | ≤ 0.005 % |
ACS - Nitrate (NO₃) | ≤ 0.003 % |
ACS - Sulfate (SO₄) | ≤ 0.01 % |
ACS - Ammonium (NH₄) | ≤ 0.005 % |
ACS - Calcium (Ca) | ≤ 0.06 % |
ACS - Iron (Fe) | ≤ 0.001 % |
ACS - Lead (Pb) | ≤ 0.005 % |
ACS - Magnesium (Mg) | ≤ 0.01 % |
ACS - Potassium (K) | ≤ 0.02 % |
ACS - Sodium (Na) | ≤ 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...