Copper(I) oxide, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
|
Warning
|
Synonyms:
Cuprous oxide
, Dicopper oxide
1878-01EA
20955
INR
1878-01
Copper(I) oxide, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Copper (I) oxide
Formula:
Cu₂O MW: 143,09 g/mol Boiling Pt: 1800 °C (1013 hPa) Melting Pt: 1232 °C Density: 5,7…6.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010974 CAS Number: 1317-39-1 EINECS: 215-270-7 UN: 3077 ADR: 9,III Merck Index: 13,02694 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (Cu₂O) | ≥95.0 % |
Preservative (zinc stearate) | 0.2 - 0.5 % |
Insoluble in HNO₃ | ≤0.5 % |
Chloride (Cl) | ≤0.5 % |
Sulfate (SO₄) | ≤0.05 % |
Iron (Fe) | ≤0.05 % |
Free Copper (Cu), For Information Only |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...