Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 1:00 am CET Saturday through 0:30 am Monday. Orders placed will be processed after maintenance is complete.

Ammonium chloride 99.5-100.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

3364-19EA 114879 INR InStock
3364-19 3364-12 3364-32
Ammonium chloride 99.5-100.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™
Ammonium chloride


Formula: NH₄Cl
MW: 53,49 g/mol
Melting Pt: 335 °C
Density: 1,5256 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
UN: 0000
Merck Index: 13,00510
https://in.vwr.com/stibo/web/std.lang.all/05/71/16640571.jpg
Order Now
Specification Test Results
Food GMP Manufactured Product
GMP Manufactured Product
Meets FCC Requirements
Meets USP Requirements
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (NH₄Cl) (Dried Basis) 99.5 - 100.5%
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Loss on Drying ≤0.5%
USP - (pH) 4.6 - 6.0
USP - Residue on Ignition ≤0.1%
USP - Thiocyanate (SCN) Passes Test
FCC - Assay (NH₄Cl) (Dried Basis) 99.5 - 100.5%
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying ≤0.5%
No Class 1,2,3 or other solvents are used/produced in the manufacturing/purification of the product
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty