AMRE0104
0104-500GEA 11533 INR
VWRC0104-500G
AMRE0104
TMA effectively abolishes the differential melting temperatures of AT and GC base pairs, making it a useful reagent for nucleic acid hybridisation procedures.