Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl ethylphosphonate ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 78-38-6
Molecular Formula: C6H15O3P
Formula Weight: 166.16
Flash Point: 105°(221°F)
Refractive Index: 1.4160
Boiling Point: 82-83°/11mm
Beilstein Reference: 1756861
UN No.: 3082
MDL No.: MFCD00039888


Formula: C₆H₁₅O₃P
MW: 166,16 g/mol
Boiling Pt: 35 °C (0,5 -rr)
Melting Pt: 197,8 °C
Density: 1,02 g/cm³ (20 °C)
MDL Number: MFCD00039888
CAS Number: 78-38-6
EINECS: 201-111-9
UN: 3082
ADR: 9,III

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g A11046.14
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Formula weight 166.16
CAS No. 78-38-6
Purity 98%
Refractive index 1.4160
Flash point 105°(221°F)
Boiling point 82-83°/11mm
Molecular formula C6H15O3P
MDL No. MFCD00039888
UN No. 3082
Beilstein reference 1756861
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty