Aluminium hydroxide, TECHNICAL
|
Warning
|
Synonyms:
Aluminum hydroxide
20980.364EA
7985
INR
20980.364
Aluminium hydroxide, TECHNICAL
Aluminium hydroxide
Formula:
Al(OH)₃ MW: 78 g/mol Melting Pt: 300 °C Density: 2,42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003420 CAS Number: 21645-51-2 EINECS: 244-492-7 Merck Index: 12,00355 |
Specification Test Results
Aluminium oxide | 60 - 70 % |
Mean particle size ( |
Max. 3 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...