Calcium fluoride ≥98%, GPR RECTAPUR®, precipitated

Warning

Synonyms: FluoriteFluorspar

22344.291EA 15084 INR
22344.291
Calcium fluoride ≥98%, GPR RECTAPUR®, precipitated
Calcium fluoride
Formula: CaF₂
MW: 78,07 g/mol
Boiling Pt: 2500 °C (1013 hPa)
Melting Pt: 1418 °C
Density: 3,18 g/cm³ (20 °C)
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now

Specification Test Results

Assay Min. 98 %
Heavy metals (as Pb) Max. 50 ppm
Cl (Chloride) Max. 0.1 %
SO4 (Sulphate) Max. 50 ppm
Fe (Iron) Max. 100 ppm
Mg (Magnesium) Max. 100 ppm
Mn (Manganese) Max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR