Sodium peroxodisulphate 1 mol/l in aqueous solution, AVS TITRINORM volumetric solution, for TOC

Danger

Synonyms: Disodium peroxodisulphatePeroxydisulphuric acid disodium saltSodium persulphate

306448.2500EA 0 INR
306448.2500
Sodium peroxodisulphate 1 mol/l in aqueous solution, AVS TITRINORM volumetric solution, for TOC
Sodium peroxodisulphate

Delivery information: This product is not available in all countries. Please check with your local VWR office or supplier.

Formula: Na₂S₂O₈
MW: 238,11 g/mol
MDL Number: MFCD00003501
CAS Number: 7775-27-1
EINECS: 231-892-1
UN: 3216
ADR: 5.1,III

Order Now

Specification Test Results

NIST traceable Confirmed
Assay (at preparation) 230 - 245 g/l

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR