α,α,α-Trifluoro-o-toluoyl chloride ≥98%

Danger

Synonyms: 2-(Trifluoromethyl)benzene-1-carbonyl chloride2-(Trifluoromethyl)benzoyl chloride

A10564.A1EA 0 INR
A10564.A1
α,α,α-Trifluoro-o-toluoyl chloride ≥98%
α,α,α-Trifluoro-o-toluoyl chloride
Formula: C₈H₄ClF₃O
MW: 208,57 g/mol
Boiling Pt: 89…90 °C (16 mmHg)
Melting Pt: –22 °C
Density: 1,414
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000667
CAS Number: 312-94-7
EINECS: 206-233-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR