3-Chlorophenyl isocyanate ≥97%

Synonyms: Isocyanic acid 3-Chlorophenyl ester

L11483.22EA 0 INR
L11483.22
3-Chlorophenyl isocyanate ≥97%
3-Chlorophenyl isocyanate
Formula: C₇H₄ClNO
MW: 153,57 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –4,4 °C
Density: 1,26 g/cm³ (20 °C)
MDL Number: MFCD00002016
CAS Number: 2909-38-8
EINECS: 220-822-5
UN: 3382
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR