1-Heptanesulphonic acid sodium salt for ion pair chromatography
Synonyms:
Sodium 1-heptanesulphonate
, Sodium heptane-1-sulphonate
20335.104EA
16017
INR
20335.104
1-Heptanesulphonic acid sodium salt for ion pair chromatography
1-Heptanesulphonic acid sodium salt
Formula:
H₃C(CH₂)₅CH₂SO₃Na MW: 202,25 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007543 CAS Number: 22767-50-6 EINECS: 245-210-5 |
Specification Test Results
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...