Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

dNTPs

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"12"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Ready to use molecular biology grade dNTP mixes and dNTP sets. The dNTP mix is designed to save hands-on time for researchers and reduce the possibili...
Catalog Number: (E542-40UM)

Supplier:  VWR Chemicals
Description:   Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA.
UOM:  1 * 40 µmol
Catalog Number: (N557-0.5ML)

Supplier:  VWR Chemicals
Description:   Ready to use dNTP mixture, 10 mM solution.
UOM:  1 * 0,5 ml
Catalog Number: (E533-40UM)

Supplier:  VWR Chemicals
Description:   dATP, 100 mM solution in sterile, deionised water at pH 7,0 to maintain stability. Prepared from monosodium salts of the free nucleotides.
UOM:  1 * 40 µmol
Catalog Number: (E636-40UMOLE)

Supplier:  VWR Chemicals
Description:   Ready to use dNTP mix. One tube of 10 μmol of each dNTP in a 25 mM solution, pH 7,0. Tube volume 400 μl.
UOM:  1 * 40 µmol
Catalog Number: (732-2785)

Supplier:  VWR Peqlab
Description:   dNTP mix contains a ready-to-use mixture of dATP, dCTP, dGTP and dTTP. Each synthetically manufactured deoxynucleotide has a concentration of 10 mM. A...
UOM:  1 * 1 items
Catalog Number: (K461-KIT)

Supplier:  VWR Chemicals
Description:   Set contains one vial of each dNTP (dATP, dTTP, dCTP, dGTP), 40 μmol of 100 mM solution, pH 7,0.
UOM:  1 * 1 KIT
Catalog Number: (E536-40UM)

Supplier:  VWR Chemicals
Description:   dCTP, 100 mM solution in sterile, deionised water at pH 7,0 to maintain stability.  Prepared from monosodium salts of the free nucleotides. No ex...
UOM:  1 * 40 µmol
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 12
  1  2  Next 

a new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping