Adipic acid ≥99%, TECHNICAL

Warning

Synonyms: 1,6-Hexanedioic acidHexanedioc acid

20115.365EA 10434 INR
20115.365
Adipic acid ≥99%, TECHNICAL
Adipic acid
Formula: HOOC(CH₂)₄COOH
MW: 146,14 g/mol
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1,36 g/cm³ (25 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 13,00163
https://in.vwr.com/stibo/web/std.lang.all/02/30/2300230.jpg

Order Now

Specification Test Results

Assay (calculated on dried substance) Min. 99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR