Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sorbic acid ≥99%, TECHNICAL

Warning

20664.293EA 8449 INR InStock
20664.293
Sorbic acid ≥99%, TECHNICAL
Sorbic acid


Formula: CH₃CH=CHCH=CHCOOH
MW: 112,13 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 132…135 °C
Density: 1,204 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
UN: 0000
Merck Index: 12,08869
https://in.vwr.com/stibo/web/std.lang.all/29/27/2302927.jpg
Order Now
Specification Test Results
Assay Min. 99 %

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty