L(+)-Arginine, BioChemica®

Supplier: PanReac AppliChem
Warning

Synonyms: H-L-Arg-OHArginine(S)-(+)-ArginineH-Arg-OH

A3675.1000EA 0 INR
A3675.1000 A3675.0500 A3675.0100
L(+)-Arginine, BioChemica®
L(+)-Arginine
Formula: C₆H₁₄N₄O₂
MW: 174,2 g/mol
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
UN: 0000
Merck Index: 13,00785

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR