Penicillin G sodium salt
Supplier: BIOWEST
Synonyms:
Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
, Benzylpenicillin sodium salt
, 6-(2-Phenylacetamido)penicillanic acid sodium salt
P0018-1MUEA
0
INR
P0018-1MU
Penicillin G sodium salt
Penicillin G sodium salt
Penicillin G sodium salt inhibits the final stages of bacteria cell wall synthesis. It inhibits primarily Gram-positive bacteria.
- Storage temperature of 2 to 8 °C
- pH of 5,5 to 7,5
- Shelf life of 36 months
Caution: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.
Formula:
C₁₆H₁₇N₂NaO₄S MW: 356,38 g/mol |
MDL Number:
MFCD00069666 CAS Number: 69-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...