DNA ladder, 1 kb, Ready Ladder
N551-1.2MLEA 12229 INR
VWRCN551-1.2ML
DNA ladder, 1 kb, Ready Ladder
Ladder with 11 fragments ranging from 500 to 10000 bp for easy band identification. Ready to use formula pre-mixed with loading dye.