Manganese(II) sulphate monohydrate, Reagent Grade
Supplier: VWR Chemicals
|
Warning
|
Synonyms:
Manganous sulphate monohydrate
, Manganese sulphate monohydrate
, Manganese(II) sulphate hydrate
0248-2.5KGEA
20819
INR
0248-2.5KG
0248-500G
Manganese(II) sulphate monohydrate, Reagent Grade
Manganese (II) sulphate monohydrate
Formula:
MnSO₄·1H₂O MW: 169,02 g/mol Melting Pt: 700 °C Density: 2,95 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00149159 CAS Number: 10034-96-5 EINECS: 232-089-9 UN: 3077 ADR: 9,III Merck Index: 13,05763 |
Specification Test Results
Heavy Metals (as Pb) | <0.05% |
Insolubles | <0.08% |
Purity | >98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...