di-Sodium tetraborate decahydrate ≥99.5% ACS
|
Danger
|
Synonyms:
Sodium tetraborate decahydrate
, Borax tetra
0390-5KGEA
21368
INR
0390-5KG
0390-1KG
di-Sodium tetraborate decahydrate ≥99.5% ACS
di-Sodium tetraborate decahydrate
Formula:
Na₂B₄O₇·10H₂O MW: 381,37 g/mol Boiling Pt: 320 °C (1013 hPa) Melting Pt: 75 °C Density: 1,73 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 EINECS: 215-540-4 REACH: 01-2119490790-32 Merck Index: 13,08662 |
Specification Test Results
Calcium | <0.005% |
Chloride | ≤0.001% |
Heavy Metals | ≤0.001% |
Insolubles | ≤0.005% |
Iron | ≤0.0005% |
pH (0.01 M, Water) @25 °C | 9.15 − 9.2 |
Phosphate | ≤0.001% |
Purity | 99.5 − 105% |
Sulphate | ≤0.005% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...