Hydrazinium sulphate

Supplier: Hach
Danger

Synonyms: Hydrazine sulphate saltHydrazine sulphate

705-0057EA 0 INR
705-0057
Hydrazinium sulphate
Hydrazinium sulphate
Formula: NH₂NH₂·H₂SO₄
MW: 130,12 g/mol
Melting Pt: 254 °C
Density: 1,37 g/cm³ (20 °C)
MDL Number: MFCD00044873
CAS Number: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1,III
Merck Index: 13,04791
https://in.vwr.com/stibo/web/std.lang.all/91/35/2299135.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR