Malachite green oxalate, high purity
Supplier: VWR Chemicals
|
Danger
|
Synonyms:
Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalate
, N-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)
, Basic Green 4
, N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate
0186-100GEA
0
INR
0186-100G
0186-250G
Malachite green oxalate, high purity
Malachite green oxalate
Formula:
C₅₂H₅₄N₄O₁₂ MW: 927,02 g/mol Melting Pt: ∼159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011766 CAS Number: 2437-29-8 EINECS: 219-441-7 UN: 2811 ADR: 6.1,III Merck Index: 13,05722 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...