L(+)-Cysteine hydrochloride monohydrate ≥99%, high purity

Warning

Synonyms: (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate(R)-(+)-Cysteine hydrochloride monohydrateCysteine Hydrochloride monohydrateH-Cys-OH.HCl.H2OCysteine Hydrochloride, Monohydrate

0206-250GEA 7564 INR
0206-250G
L(+)-Cysteine hydrochloride monohydrate ≥99%, high purity
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175,64 g/mol
Melting Pt: 168 –170 °C
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://in.vwr.com/stibo/web/std.lang.all/58/37/2345837.jpg

Order Now

Specification Test Results

Arsenic <0.00015%
Chloride Content 19.8 – 20.8%
Heavy Metals (as Pb) <0.0015%
Iron <0.003%
Loss on Drying 8 – 12%
Purity >99%
Residue on Ignition <0.4%
Specific Rotation +5.7 to +6.8 °
Sulphate <0.03%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR