Ammonia solution 10%
|
Danger
|
Synonyms:
Ammonium hydroxide
799384AEA
0
INR
799384A
Ammonia solution 10%
Ammonia solution
Formula:
NH₃ MW: 35,05 g/mol Density: 0,92 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8,III |
Specification Test Results
Ammonia (W/V) | 9.5 - 10.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...