Sulphuric acid ≥85%

Supplier: VWR Chemicals
Danger

799379SEA 0 INR
799379S
Sulphuric acid ≥85%
Sulphuric acid
Formula: H₂SO₄
MW: 98,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Assay (H2SO4) 84.5 - 85.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR