Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium permanganate standardised solution 0.1 N in aqueous solution

Supplier: Alfa Aesar

CAS No.: 7722-64-7
Molecular Formula: KMnO4
Formula Weight: 158.03
Physical Form: Liquid
Sensitivity: Light Sensitive
Appearance: Dark purple
MDL No.: MFCD00011364


Formula: KMnO₄
MW: 158,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 1 l 35590.K2
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Sensitivity Light Sensitive
Formula weight 158.03
CAS No. 7722-64-7
Physical form Liquid
Molecular formula KMnO4
Appearance Dark purple
MDL No. MFCD00011364
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty