Tween® 80 (Polysorbate), viscous liquid, Proteomics Grade
Supplier: VWR Chemicals
M126-1LEA
7649
INR
M126-1L
M126-100ML
Tween® 80 (Polysorbate), viscous liquid, Proteomics Grade
Tween® 80 (Polysorbate)
Non-ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.
- High purity and protease-free
- Suitable for most biological applications
Non-ionic detergents have uncharged hydrophilic groups and are gentle, non-denaturing detergents ideal for studying the conformation and function of proteins, because they keep secondary and tertiary structure intact.
Boiling Pt:
>100 °C (1013hPa) Melting Pt: -21 °C Flash Pt: 149 °C (closed cup) |
MDL Number:
MFCD00082107 CAS Number: 9005-65-6 UN: 0000 Merck Index: 13,07664 |
Specification Test Results
DNase | None detected |
Heavy Metals | < 0,001 % |
Hydroxyl Number | 65 - 80 |
Identification | Passes |
Moisture (KF) | ≤3,0 % |
Protease | None detected |
Residue on Ignition | ≤0,25 % |
RNase | None detected |
Specific Gravity | 1,06 - 1,09 |
Viscosity | 300 - 500 cS |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...