TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) ≥98.0% for molecular biology


Warning

Synonyms: (p-tert-Octylphenoxy)polyethoxyethanolO-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanolOctoxynolPolyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether

437002AEA 4375 INR
437002A
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) ≥98.0% for molecular biology
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Non-ionic surfactant, suitable for use in molecular biology applications.


Triton® is a registered trade mark of Union Carbide

Boiling Pt: >200 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1,06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III
Merck Index: 13,06793

Order Now

Specification Test Results

Assay Min. 98.0 %
Appearance Clear viscous liquid
Identity (IR) Passes test
DNases Not detected
RNases Not detected
Proteases Not detected
Density (20/4) 1.0640 - 1.0670
pH (1 %) 6.00 - 8.00
Water Max. 1.0 %
Heavy metals (as Pb) Max. 0.0005 %
SO4 (Sulphate) Max. 0.005 %
Cl (Chloride) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR