Polyethylene glycol 1,500, TECHNICAL
26604.296EA
3681
INR
26604.296
Polyethylene glycol 1,500, TECHNICAL
Polyethylene glycol 1,500
Formula:
H(OCH₂CH₂)nOH MW: 1500 g/mol Boiling Pt: >250 °C (1013 hPa) Melting Pt: 44…48 °C Density: 1,2 g/cm³ (20 °C) Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Specification Test Results
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...