Streptomycin sulphate USP
Supplier: VWR Chemicals
|
Warning
|
0382-500GEA
0
INR
0382-500G
0382-100G
Streptomycin sulphate USP
Streptomycin sulphate
30 μg/ml, binds to the 30S subunit of bacterial ribosome.
Formula:
C₂₁H₃₉N₇O₁₂·1.5H₂SO₄ MW: 1457,4 g/mol Melting Pt: 250 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037023 CAS Number: 3810-74-0 EINECS: 223-286-0 UN: 2811 ADR: 6.1,III Merck Index: 12,08983 |
Specification Test Results
Expiration Date | Report |
Identification | Pass |
Loss on Drying (USP) | 5.0% |
pH (20%, Water) at 25 °C | 4.5 - 7.0 |
Potency (as is) | 0 - 9999 μg/mg |
Potency (Dry Basis) | 650 - 850 µg/mg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...