Neomycin sulphate for tissue culture, γ-irradiated
Synonyms:
Fradiomycin sulphate
, Neomycin
, Neomycin sulphate salt
, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate
Aseptic, gamma-irradiated tissue culture tested neomycin sulfate preparation. Inhibits protein synthesis in gram-positive and gram-negative bacteria. Selection agent used to establish stable mammalian cell lines.
When reconstituted in 20ml sterile water contains 10 mg neomycin per ml.
Formula:
C₂₃H₄₆N₆O₁₃·3H₂SO₄ MW: 908,87 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150827 CAS Number: 1405-10-3 EINECS: 215-773-1 Merck Index: 12,06542 |
Specification Test Results
pH (3.3%, Water) @25°C | 5.0 - 7.5 |
Potency (anhydrous) |
|
Solubility (Vial Contents, 20ml Water) | Pass |
Sterile (Gamma-Irradiated) | Pass |
USP Grade Neomycin Sulfate | Pass |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...