Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D(+)-Mannose ≥99.5%, high purity

Supplier: VWR Chemicals



Formula: C₆H₁₂O₆
MW: 180,16 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 133 °C
Density: 1,539 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064122
CAS Number: 3458-28-4
EINECS: 222-392-4
UN: 0000
Merck Index: 13,05772
Specification Test Results
Loss on Drying < 1%
Melting Point 125°C - 131°C
Purity (HPLC) > 99.5 %
Solubility (5%, Water) Pass
Specific Rotation 14° - 15°

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle for solids 25 g J443-25G
Plastic bottle for solids 100 g J443-100G
Plastic bottle for solids 500 g J443-500G
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty