Cyto-paraffin wax, Solidification point: 54 - 58°C Contains 0,05% of yellow beeswax, bar, MICROCOLOR® for histology

Synonyms: Wax ParaffinParaffin wax

26177.290EA 12807.9 INR
26177.290
Cyto-paraffin wax, Solidification point: 54 - 58°C Contains 0,05% of yellow beeswax, bar, MICROCOLOR® for histology
Paraffin

Contains 0,05% of yellow beeswax

Melting Pt: 54…58 °C
Density: 0,9 g/cm³ (20 °C)
Flash Pt: >100 °C (closed cup)
CAS Number: 8002-74-2
EINECS: 232-315-6

Order Now

Specification Test Results

Identification Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR