Magnesium ≥99.8%, turnings, Technical Grade
|
Danger
|
25073.237EA
5879
INR
25073.237
Magnesium ≥99.8%, turnings, Technical Grade
Magnesium
Formula:
Mg MW: 24,3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1,III |
Specification Test Results
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...