Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "tri-sodium-citrate-dihydrate"

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   tri-Sodium citrate dihydrate AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Description:   tri-Sodium citrate dihydrate, reagent grade
Supplier:  VWR Chemicals
Description:   tri-Sodium citrate molecular biology grade 1 * 100 g
Supplier:  VWR Chemicals
Description:   Tri-sodium citrate dihydrate crystallized ph. eur. 1 * 25 kg
UOM:  1 * 25 kg
Supplier:  VWR Chemicals
Description:   tri-Sodium citrate dihydrate GPR RECTAPUR®
UOM:  1 * 25 kg
Catalog Number: (HACH1490849)
Supplier:  Hach
Description:   SODIUM CITRATE FOR SILICA 1 * 500 ml
UOM:  1 * 500 ml
Catalog Number: (BDHB0288-2.5KG)
Supplier:  BDH (US PRODUCED)
Description:   tri-Sodium citrate dihydrate
UOM:  1 * 2,5 kg
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7  of 7