Paraformaldehyde
Supplier:
VWR Chemicals
Description:
PARAFORMALDEHYDE PRILLS (91-93%) REAGENT GRADE 1 * 500 g
Catalog Number:
(28794.460)
Supplier:
VWR Chemicals
Description:
Paraformaldehyde, powder TECHNICAL
UOM:
1 * 25 kg
Supplier:
Alfa Aesar
Description:
Paraformaldehyde, 16% w/v aq. soln., methanol free 1 * 10 items
Catalog Number:
(J61899.AP)
Supplier:
Alfa Aesar
Description:
Paraformaldehyde, 4% in PBS 1 * 500 ml
UOM:
1 * 500 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
|
|||||||||