You Searched For: Ammonium thiocyanate


7  results were found

SearchResultCount:"7"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR Chemicals
Description: Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Description: Ready to use test solution (TS).
Supplier: VWR Chemicals
Description: Ammonium thiocyanate ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Catalog Number: (5627-02)
Supplier: Avantor
Description: Standardised at 25 °C against chemicals whose certification is traceable to NIST. Lot standardisation value, accurate to ±1 part per 1000, provided on...
UOM: 1 * 1 L

Catalog Number: (21342.297)
Supplier: VWR Chemicals
Description: Ammonium thiocyanate ≥98%, purified
UOM: 1 * 1 kg

Catalog Number: (31960.291)
Supplier: VWR Chemicals
Description: Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
UOM: 1 * 1 L

Catalog Number: (87775.290)
Supplier: VWR Chemicals
Description: Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1006701
UOM: 1 * 1 L

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7 of 7
no targeter for Bottom