You Searched For: di-Ammonium hydrogen orthophosphate


4  results were found

SearchResultCount:"4"

Sort Results

List View Easy View

Rate These Search Results

Supplier: Avantor
Description: Diammonium hydrogen orthophosphate ≥98.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: VWR Chemicals
Description: Diammonium hydrogen orthophosphate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Catalog Number: (21306.362)
Supplier: VWR Chemicals
Description: Diammonium hydrogen orthophosphate ≥97.5%, TECHNICAL
UOM: 1 * 5 kg

Supplier: VWR Chemicals
Description: Diammonium hydrogen orthophosphate ≥98% ACS
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 4 of 4
no targeter for Bottom