Thermo-hygrometer with probe, Traceable®
VWRU35519-048EA 0 INR
VWRU35519-048
Thermo-hygrometer with probe, Traceable®
This thermo-hygrometer with temperature probe continuosly monitors and measures temperature and humidity.
  • Simultaneously displays both temperature and relative humidity on dual-display LCD (13 mm character height)
  • Minimum/maximum memory
  • Interface for transfer of data to a computer
  • Always-on instrument (no off switch) makes continuous readings
Certifications: Compliant ISO/IEC 17025, ANSI/NSCL Z540-1 and 9001.
Traceable to NIST standards.
Delivery information: Supplied with temperature probe and 3,01 m cable, Traceable® certificate of calibration, hard plastic carrying case and two AA alkaline battery.