VWR®, Undercounter Laboratory Refrigerator
Supplier: VWR
VWR®
VWRU89031-974EA 0 INR
VWRU89031-974
VWR®, Undercounter Laboratory Refrigerator
Refrigerators
This laboratory refrigerator unit is designed to meet the demanding standards of clinical, life science, pharmaceutical, biotechnology and industrial applications.
  • Exceptional temperature control and uniformity
  • Microprocessor control with user-programmable functions and features
  • Built-in front-breather condenser
Units offer exceptional temperature control and uniformity via a door-mounted microprocessor controller and interior forced air circulation. The easy-to-read controller features an upward angled LED display and keypad. Digital temperature input allows end user temperature flexibility. The controller lockout function prevents set point tampering.
A pre-set alarm at ±3 °C from temperature set point monitors critical variances. A door-ajar alarm with alarm delay timer eliminates nuisance alarms and remote alarm contacts allow for connection to a centralised alarm monitoring system.
A built-in front-breather condenser allows units to be placed under benches or countertops and requires no additional clearance from the top, back or sides of the cabinet. These compact units can also be left free standing, placed side by side or securely stacked to create a combination refrigerator/freezer.
A keyed door ensures safe and secure storage within each unit. The refrigerator is equipped with adjustable wire shelves, the freezer is equipped with fixed evaporator shelves and both units feature a right-hinged door swing and a rounded door profile and handle.
Certifications: Meets JCAHO standards for controlling medication access.
Ordering information: A standard access port (30 mm) and stacking kits are available separately.