Series glass door chromatography refrigerators with natural refrigerants
VWRU10819-302EA 773382 INR
VWRU10819-306 VWRU10819-300 VWRU10819-310 VWRU10819-298 VWRU10819-302
Series glass door chromatography refrigerators with natural refrigerants
Units feature a digital microprocessor temperature controller, allowing for the precise temperature management necessary for critical samples and supplies.
  • Adjustable operating temperature range: 1 to 10 °C
  • Sample simulated digital temperature display
  • Audible and visual high/low temperature alarms
  • Swivel casters add four inches to height
  • Interior duplex electrical outlet
  • Keyed door locks
A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings. An automatic cycle-defrost design prevents product damaging temperature spikes.
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates, as well as UL, ASHRAE compliance guidelines.
Certifications: UL/C-UL Listed.
Delivery information: Units are supplied with a two-years parts and labour warranty, with an additional five-years warranty on compressor parts.