Gel imaging systems, GelStudio PLUS and GelStudio PLUS touch
Supplier: UVP
UVPA849-00552-2EA 0 INR
UVPA849-00552-2 UVPA849-00553-2
Gel imaging systems, GelStudio PLUS and GelStudio PLUS touch
The UVP® GelStudio PLUS and UVP® GelStudio PLUS touch systems are essential to laboratories performing genomic and proteomic applications. These gel documentation systems are used for high sensitivity imaging of DNA gels, protein gels, Coomassie Blue, GelGreen, SYBR Green and a wide range of genomic applications. The ergonomically designed GelStudio PLUS is available as a stand-alone system as well as an external computer version for those laboratories with particular IT/computer restrictions.
  • High resolution 5,0 megapixel camera, with an automated f/1,2 zoom lens and 16-bit file depth
  • Integrated UV protection shield, maximises protection from UV radiation while working over the transilluminated surface
  • Wide maximum illuminated imaging area for imaging multiple gels of various sizes (up to 250×260 mm)
Darkroom features a thin-line transilluminator in a slide-out tray for easy access, with uniform and high-intensity UV lighting in a compact form designed with long-life UV lamps, for minimum service requirements "Slide2Hide" door, features smooth operation and limits bench top interferences
Overhead white lighting is included, with optional UV light. Five-position automated, filter tray for easy access (ethidium bromide filter included)
Integrated, 15.6 inch, multi-touch computer, with large storage capacity offers fully networkable system that can connect over wireless network for simple transfer of files and data (GelStudio PLUS touch only)
Delivery information: All systems include multiple copies of the full package VisionWorks® Acquisition and Analysis Software for use on an external computer. VisionWorks® touch software is included with the stand-alone system only.
Also included are a thin-line 302 nm UV transilluminator, filter size 250×260 mm, HIGH/LOW intensity switch; ethidium bromide filter; slide-down UV protection shield; fluorescent target; gel tray; gel ruler; and empty flash drive for data and image storage. GelStudio PLUS touch also supplied with keyboard and mouse.
Additional filters and lighting options are available separately. Please contact your local VWR sales office for further details.