VWR® Symphony™ 4417R, Centrifuges, bench top, refrigerated
Supplier: VWR
VWR®
VWRU89176-486EA 0 INR
VWRU89176-496 VWRU89176-490 VWRU89176-486
VWR® Symphony™ 4417R, Centrifuges, bench top, refrigerated
Centrifuges Microcentrifuges
These universal table-top centrifuges are ideal for use with all blood collection tubes and microplates.
  • All relevant run parameters can easily be set with display keys and be pre-selected with the main adjustment knob
  • All pre-selected and actual values are permanently displayed on the large LCD display
  • Lid is latched and released with an electromagnetic lid lock
  • Powerful, maintenance-free brushless induction drive with a low noise level
  • Rotor recognition provides safety and convenience when different rotors are used
Various fixed angle and swing-out rotors are available to process a variety of different tube types from 0,2 to 100 ml. These units are well suited for all standard blood collection, conical and microcentrifuge tubes, as well as standard and deep-well microplates. High speed and g-force make the 4417 series ideal for applications ranging from clinical to cell culture and molecular biology. Powerful, brushless motors quickly accelerate rotors to the preset speed. Braking is dynamically controlled to bring the rotors to a soft stop without disrupting fragile samples. Operation of the centrifuge may be timed, continuous or momentary. Each rotor is recognized at the beginning of a run and automatically limited to its rated maximum speed. In addition, by recognising the rotor, the microprocessor can accurately calculate and display g-force during a run.
Ordering information: Rotors need to be ordered separately. Cell culture bundles are supplied with rotor.